missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COPG Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
€ 280.00
Specifications
| Antigen | COPG |
|---|---|
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
COPG Polyclonal specifically detects COPG in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| COPG | |
| Polyclonal | |
| Rabbit | |
| Human | |
| coat protein gamma-cop, coatomer protein complex, subunit gamma, coatomer protein complex, subunit gamma 1, COPG1coatomer subunit gamma, FLJ21068, Gamma-coat protein, gamma-COP | |
| COPG1 | |
| IgG | |
| Affinity Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 22820 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:MLPSILVLLKRCVMDDDNEVRDRATFYLNVLEQKQKALNAGYILNGLTVSIPGLERALQQYTLEPSEKPFDLKSVPL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title