Learn More
Hach Lange™ Copper Cuvette Tests
Brand:  Hach Lange™ LCK329
Additional Details : Weight : 0.50000kg
Product Code. 10402353
  • € 160.00 / Each
Estimated Shipment: 02-05-2024
to see stock.
Add to basket
Description
Description
Order Info
dimRange
Specifications
Specifications
25
15°C to 25°C
Copper Cuvette Test
0.1 to 8.0mg/L Cu
Bathocuproine Disulphonic Acid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers