missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) 2,4-pentanedionate hydrate, Thermo Scientific Chemicals

Catalog Number p-7049027
16.00 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
13395-16-9
C10H14CuO4
261.76
MFCD00000016
QYJPSWYYEKYVEJ-FDGPNNRMSA-L
Bis(acetylacetonato)copper(II); Bis(2,4-pentanedionato)copper(II)
copper(2+) bis((2Z)-4-oxopent-2-en-2-olate)
[Cu++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
13395-16-9
C10H14CuO4
261.76
MFCD00000016
QYJPSWYYEKYVEJ-FDGPNNRMSA-L
Bis(acetylacetonato)copper(II); Bis(2,4-pentanedionato)copper(II)
copper(2+) bis((2Z)-4-oxopent-2-en-2-olate)
[Cu++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Copper(II) acetylacetonate catalyzes coupling and carbene transfer reactions. Metal acetylacetonates are used as catalysts for polymerization of olefins and transesterification. They are used as PVC stabilizer. They are used as curing agents for epoxy resins, acrylic adhesives and silicone rubbers. They are used as solvents, lubricant additives, paint drier, and pesticides. They are used in glass coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 230°C
Odor Weak
Quantity 25 g
Solubility Information Slightly soluble in water. Soluble in alcohol.
Formula Weight 261.76 (Anhydrous)
Physical Form Powder
Chemical Name or Material Copper(II) 2,4-pentanedionate hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'