missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) bromide, 99%, for analysis, anhydrous, Thermo Scientific Chemicals

Catalog Number 15259886
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7789-45-9
Br2Cu
223.36
Cupric bromide
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7789-45-9
Br2Cu
223.36
Cupric bromide

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 900.0°C
Melting Point 498.0°C
Quantity 1 kg
Assay Percent Range 98.5% min. (Iodometry)
Linear Formula CuBr2
Merck Index 15,2619
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, acetone, nh3, practically insoluble in benzene, diethyl ether,, conc. h2so4
Formula Weight 223.36
Percent Purity 99%
Grade Analytical
Loss on Drying 1% max.
Chemical Name or Material Copper(II) bromide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Copper(II) bromide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'