missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) gluconate, Thermo Scientific Chemicals

Catalog Number p-7049591
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy
527-09-3
C12H22CuO14
453.84
MFCD00075297
OCUCCJIRFHNWBP-IYEMJOQQSA-L
copper gluconate
131854750
CHEBI:31431
copper;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
C(C(C(C(C(C(=O)[O-])O)O)O)O)O.C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Cu+2]
This item is not returnable. View return policy
527-09-3
C12H22CuO14
453.84
MFCD00075297
OCUCCJIRFHNWBP-IYEMJOQQSA-L
copper gluconate
131854750
CHEBI:31431
copper;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
C(C(C(C(C(C(=O)[O-])O)O)O)O)O.C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Cu+2]

Copper(II) gluconate is used in dietary supplement. It is also used to treat acne vulgaris, common cold, hypertension, premature labor, leishmaniasis and visceral postoperative complications. Further, it is used in oral deodorants and feed additives. In addition to this, it is used as a synergist and nutritional supplement for foods. It is the active component in retsyn used in mints and certs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 155°C to 157°C
Quantity 50 g
Merck Index 14,2640
Solubility Information Soluble in water. Insoluble in acetone,alcohol and ether.
Formula Weight 453.85
Physical Form Powder
Chemical Name or Material Copper(II) gluconate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'