Learn More
Bochem™ Copper/Plastic Funnel Filter
Specifically sized and suited for use in a funnel. Bochem™ Copper/Plastic Funnel Filter is made of two materials for durability and resistance.
Brand:  Bochem™ 8853
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 10058909
  • € 13.65 / Each
Estimated Shipment: 21-05-2024
to see stock.
Add to basket
Description
Description
  • Copper and plastic construction.
  • Resistant and durable.
  • Comes in two sizes.
Specifications
Specifications
Plastic
105mm
Funnel