missing translation for 'onlineSavingsMsg'
Learn More

Coppersensor 1, TRC

Product Code. 30339447
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30339447 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30339447 Supplier Toronto Research Chemicals Supplier No. TRCB874740100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Coppersensor 1
CAS 874748-20-6
Molecular Formula C30H50BF2N3S4
Quantity 100 mg
Synonym [4-Ethyl-beta-(4-ethyl-3,5-dimethyl-2H-pyrrol-2-ylidene-kN)-N,N-bis[2-[[2-(ethylthio)ethyl]thio]ethyl]-3,5-dimethyl-1H-pyrrole-2-ethanaminato-kN1]difluoro- (T-4)- Boron, (T-4)-[4-Ethyl-beta-(4-ethyl-3,5-dimethyl-2H-pyrrol-2-ylidene-kN)-N,N-bis[2-[[2-(ethylthio)ethyl]thio]ethyl]-3,5-dimethyl-1H-pyrrole-2-ethanaminato-kN1]difluoroboron, CS 1
InChI Formula InChI=1S/C30H50BF2N3S4/c1-9-26-22(5)29-28(21-34(13-15-39-19-17-37-11-3)14-16-40-20-18-38-12-4)30-23(6)27(10-2)25(8)36(30)31(32,33)35(29)24(26)7/h9-21H2,1-8H3
SMILES [F-][B+3]1([F-])N2=C(C)C(=C(C)C2=C(CN(CCSCCSCC)CCSCCSCC)C3=C(C)C(=C(C)[N-]13)CC)CC
IUPAC Name N-[(5,11-diethyl-2,2-difluoro-4,6,10,12-tetramethyl-3-aza-1-azonia-2-boranuidatricyclo[7.3.0.03,7]dodeca-1(12),4,6,8,10-pentaen-8-yl)methyl]-2-(2-ethylsulfanylethylsulfanyl)-N-[2-(2-ethylsulfanylethylsulfanyl)ethyl]ethanamine
Molecular Weight (g/mol) 629.81
Formula Weight 629.2949
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.