Learn More
Coproporphyrin I Tetramethyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | Coproporphyrin I Tetramethyl Ester |
| CAS | 25767-20-8 |
| Molecular Formula | C40H46N4O8 |
| Quantity | 1 mg |
| Synonym | 3,8,13,18-Tetramethyl-21H,23H-porphine-2,7,12,17-tetrapropanoic Acid 2,7,12,17-Tetramethyl Ester, 3,8,13,18-Tetramethyl-21H,23H-porphine-2,7,12,17-tetrapropanoic Acid Tetramethyl Ester |
| InChI Formula | InChI=1S/C40H46N4O8/c1-21-25(9-13-37(45)49-5)33-18-30-23(3)27(11-15-39(47)51-7)35(43-30)20-32-24(4)28(12-16-40(48)52-8)36(44-32)19-31-22(2)26(10-14-38(46)50-6)34(42-31)17-29(21)41-33/h17-20,41,44H,9-16H2,1-8H3/b29-17-,30-18-,31-19-,32-20-,33-18-,34-17-,35-20-,36-19- |
| SMILES | COC(=O)CCC1=C(C)c2cc3nc(cc4[nH]c(cc5nc(cc1[nH]2)c(C)c5CCC(=O)OC)c(C)c4CCC(=O)OC)c(C)c3CCC(=O)OC |
| IUPAC Name | methyl 3-[(1Z,4Z,9Z,15Z)-7,12,17-tris(3-methoxy-3-oxopropyl)-3,8,13,18-tetramethyl-21,23-dihydroporphyrin-2-yl]propanoate |
| Molecular Weight (g/mol) | 710.82 |
| Formula Weight | 710.3316 |