missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COPS3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111318
This item is not returnable.
View return policy
Description
Immunogen sequence: LRGIGILKQA IDKMQMNTNQ LTSIHADLCQ LCLLAKCFKP ALPYLDVDMM DICKENGAYD AKHFLCYYYY GGMIYTGLKN FERALYFYEQ AITTPAM Highest antigen sequence indentity to the following orthologs: Rat - 99%, Mouse - 99%.
The protein encoded by this gene possesses kinase activity that phosphorylates regulators involved in signal transduction. It phosphorylates I kappa-Balpha, p105, and c-Jun. It acts as a docking site for complex-mediated phosphorylation. The gene is located within the Smith-Magenis syndrome region on chromosome 17.
Specifications
| COPS3 | |
| Polyclonal | |
| Unconjugated | |
| Cops3 | |
| COP9 (constitutive photomorphogenic) homolog, subunit 3; COP9 (constitutive photomorphogenic) homolog, subunit 3 (Arabidopsis thaliana); COP9 (constitutive photomorphogenic), subunit 3; COP9 complex S3; COP9 complex subunit 3; COP9 constitutive photomorphogenic homolog subunit 3; COP9 signalosome complex subunit 3; COP9 signalosome subunit 3; COPS 3; COPS3; Csn3; JAB1-containing signalosome subunit 3; SGN3; signalosome subunit 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8533 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UNS2 | |
| Cops3 | |
| Recombinant Protein corresonding to Human COPS3. Recombinant protein control fragment (Product #RP-104907). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction