Learn More
Invitrogen™ COPS3 Polyclonal Antibody

Description
Immunogen sequence: GLKNFERALY FYEQAITTPA MAVSHIMLES YKKYILVSLI LLGKVQQLPK YTSQIVGRFI KPLSNAYHEL AQVYSTNNPS ELRNLVNKHS ETFTRDNNMG LVKQCLSSLY KKNIQRLTKT FLTLSLQDMA SRVQLSGPQE AEKYVLHMIE DGEIFASINQ KDGMVSFHDN PEKYNNPAML HNIDQEMLKC IELDERLKAM DQEITVNPQF VQKSMGSQED DSGNKPSSYS; Positive Samples: SW480, HeLa, 293T, Mouse heart, Rat heart, Rat brain; Cellular Location: Cytoplasm, Nucleus.
Specifications
Specifications
| Antigen | COPS3 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 3.41 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Cops3 |
| Gene Accession No. | O88543, Q68FW9, Q9UNS2 |
| Gene Alias | COP9 (constitutive photomorphogenic) homolog, subunit 3; COP9 (constitutive photomorphogenic) homolog, subunit 3 (Arabidopsis thaliana); COP9 (constitutive photomorphogenic), subunit 3; COP9 complex S3; COP9 complex subunit 3; COP9 constitutive photomorphogenic homolog subunit 3; COP9 signalosome complex subunit 3; COP9 signalosome subunit 3; COPS 3; COPS3; Csn3; JAB1-containing signalosome subunit 3; SGN3; signalosome subunit 3 |
| Gene Symbols | Cops3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.