missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COPS3 Polyclonal Antibody

COPS3 Polyclonal Antibody
Brand: Invitrogen PA599067
This item is not returnable.
View return policy
Description
COPS3 Polyclonal Antibody for Western Blot, IHC (P), IP, ELISA
Specifications
| COPS3 | |
| Polyclonal | |
| Unconjugated | |
| Cops3 | |
| COP9 (constitutive photomorphogenic) homolog, subunit 3; COP9 (constitutive photomorphogenic) homolog, subunit 3 (Arabidopsis thaliana); COP9 (constitutive photomorphogenic), subunit 3; COP9 complex S3; COP9 complex subunit 3; COP9 constitutive photomorphogenic homolog subunit 3; COP9 signalosome complex subunit 3; COP9 signalosome subunit 3; COPS 3; COPS3; Csn3; JAB1-containing signalosome subunit 3; SGN3; signalosome subunit 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26572, 8533 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O88543, Q9UNS2 | |
| Cops3 | |
| Recombinant Human COP9 signalosome complex subunit 3 protein (194-423AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction