Learn More
COPS8/COP9 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10089-2-AP-150UL
 View more versions of this product
Product Code. 16866243
  • € 410.00 / 150µL
Description
COPS8 is a COP9 signalosome (CSN), which is an evolutionarily conserved protein complex of the eight subunits that interacts with deubiquitinating enzymes and protein kinases and is highly homologous to the lid sub-complex of 26S proteasome. The CSN complex is an essential regulator of the ubiquitin conjugation pathway by mediating the deneddylation of the SCF-type E3 ligase complexes, which leads to a decrease in ubiquitin ligase activity of SCF-complexes such as SCF, CSA or DDB2. It is also involved in phosphorylation of p53, c-jun/JUN, ITPK1 and IRF8/ICSBP, possibly via its association with CK2 and PKD kinases. CSN8 encodes the smallest and the least conserved but first identified subunit of CSN. Recent studies show CSN8 is essential for Drosophila development and is essential for peripheral T cell homeostasis and antigen receptor-induced entry into the cell cycle from quiescence.
Specifications
Specifications
COPS8/COP9
Polyclonal
Unconjugated
COPS8
COP9, COP9 homolog, COPS8, CSN8, hCOP9, SGN8, Signalosome subunit 8
Rabbit
Antigen Affinity Chromatography
RUO
108679, 10920
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8VBV7, Q99627
Cops8
COPS8/COP9 Fusion Protein Ag0143
150 μL
Primary
Human, Mouse