missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COQ4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 415.00 - € 572.00
Specifications
| Antigen | COQ4 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18217333
|
Novus Biologicals
NBP2-58147 |
100 μL |
€ 572.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18679016
|
Novus Biologicals
NBP2-58147-25ul |
25 μL |
€ 415.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
COQ4 Polyclonal specifically detects COQ4 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| COQ4 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 51117 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:FEAVQTGLPMCILGAFFGPIRLGAQSLQVLVSELIPWAVQNGRRAPCVLNLYYERRWEQSLRALREELGITAPPMHVQGLA | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| CGI-92, Coenzyme Q biosynthesis protein 4 homolog, coenzyme Q4 homolog (S. cerevisiae), coenzyme Q4 homolog (yeast), ubiquinone biosynthesis protein COQ4 homolog, mitochondrial | |
| COQ4 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title