missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COQ7 Antibody (06), Alexa Fluor™ 594 , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-05806AF594
This item is not returnable.
View return policy
Description
COQ7 Monoclonal antibody specifically detects COQ7 in Human samples. It is validated for ELISA, ELISA
Specifications
| COQ7 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| CAT5, CLK1, CLK-1, Coenzyme Q biosynthesis protein 7 homolog, coenzyme Q, 7 (rat, yeast) homolog, coenzyme Q7 homolog, ubiquinone (yeast), COQ7 coenzyme Q, 7 homolog ubiquinone, placental protein KG-20, Timing protein clk-1 homolog, ubiquinone biosynthesis protein COQ7 homolog | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human COQ7 (Q99807-1; Ser37-Leu217). | |
| 0.1 mL | |
| Stem Cell Markers | |
| 10229 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 06 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction