missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COQ9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 216.00
Specifications
| Antigen | COQ9 |
|---|---|
| Dilution | Western Blot 1:500-1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18601576
|
Novus Biologicals
NBP2-99187-100ul |
100 μg |
N/A
|
N/A | |||||
|
18619845
|
Novus Biologicals
NBP2-99187-50ul |
50 μg |
€ 216.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
COQ9 Polyclonal antibody specifically detects COQ9 in Human samples. It is validated for Western BlotSpecifications
| COQ9 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Endocrinology, Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 57017 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| C16orf49DKFZp434K046, chromosome 16 open reading frame 49, coenzyme Q9 homolog (S. cerevisiae), coenzyme Q9 homolog (yeast), DKFZP434K046, ubiquinone biosynthesis protein COQ9, mitochondrial | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human COQ9. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title