missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COQ9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14874-1-AP-150UL
This item is not returnable.
View return policy
Description
Coenzyme Q10 (CoQ10), or ubiquinone, is a mobile lipophilic electron carrier critical for electron transfer by the mitochondrial inner membrane respiratory chain. COQ9 is 1 of several enzymes involved in biosynthesis of CoQ10 and likely functions in modification of the benzoquinone ring.Specifications
| COQ9 | |
| Polyclonal | |
| Unconjugated | |
| COQ9 | |
| C16orf49, COQ9, PSEC0129 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 498909, 57017, 67914 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75208, Q68FT1, Q8K1Z0 | |
| Coq9 | |
| COQ9 Fusion Protein Ag6645 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction