missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 405 Anti-Human CD14 (UCHM-1), Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL405-65056-100TESTS
This item is not returnable.
View return policy
Description
The CD14 antibody from Proteintech is a mouse monoclonal antibody generated with thymocytes and peripheral blood lymphocytes from a remission aml patient of human CD14. This antibody recognizes human antigen. The CD14 antibody has been validated for the following applications: FC analysis. This antibody is coralite™ plus 405.Specifications
| CD14 | |
| Monoclonal | |
| 5 μl/test | |
| Mouse | |
| Affinity purification | |
| RUO | |
| 929 | |
| Store at 2-8°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Flow Cytometry | |
| UCHM-1 | |
| CoraLite Plus 405 | |
| Thymocytes and peripheral blood lymphocytes from a remission AML patient | |
| 100 Tests | |
| Primary | |
| Human | |
| Flow Antibody | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction