missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488 Anti-Human STRO-1 (STRO-1), Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-65184-100TESTS
This item is not returnable.
View return policy
Description
The STRO-1 antibody from Proteintech is a mouse monoclonal antibody generated with human cd34 positive bone marrow cells of human STRO-1. This antibody recognizes human antigen. The STRO-1 antibody has been validated for the following applications: FC analysis. This antibody is coralite™ plus 488.Specifications
| STRO-1 | |
| Monoclonal | |
| 5 μl/test | |
| Mouse | |
| Purified by precipitation and chromatography | |
| RUO | |
| Human | |
| Flow Antibody | |
| IgM λ |
| Flow Cytometry | |
| STRO-1 | |
| CoraLite Plus 488 | |
| Human CD34 positive bone marrow cells | |
| 100 Tests | |
| Primary | |
| Store at 2-8°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction