missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated AFM Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66113-20UL
This item is not returnable.
View return policy
Description
The AFM antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human AFM. This antibody recognizes human, mouse antigen. The AFM antibody has been validated for the following applications: IF/ICC analysis. This antibody is coralite™ plus 488.Specifications
| AFM | |
| Monoclonal | |
| 1000 μg/mL | |
| ALB2; ALBA; ALF; MGC125338; MGC125339 | |
| Fusion Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| 85 kDa | |
| Liquid |
| Immunocytochemistry/Immunofluorescence | |
| 2G9F6 | |
| CoraLite Plus 488 | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 173 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction