missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated ARL13B Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66739-20UL
This item is not returnable.
View return policy
Description
The ARL13B antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ARL13B. This antibody recognizes human, canine antigen. The ARL13B antibody has been validated for the following applications: IF/ICC analysis. This antibody is coralite™ plus 488.Specifications
| ARL13B | |
| Monoclonal | |
| 1000 μg/mL | |
| ARL2L1; DKFZp686E2075; DKFZp686L2472; DKFZp686M2074; DKFZp761H079; JBTS8; MGC120611; MGC120612 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Canine | |
| 60 kDa | |
| Liquid |
| Immunocytochemistry/Immunofluorescence | |
| 1H6C3 | |
| CoraLite Plus 488 | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 200894 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction