missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated C1qC Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66268-20UL
This item is not returnable.
View return policy
Description
The C1qC antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human C1qC. This antibody recognizes human antigen. The C1qC antibody has been validated for the following applications: IF/ICC analysis. This antibody is coralite™ plus 488.Specifications
| C1qC | |
| Monoclonal | |
| 1000 μg/mL | |
| C1Q-C; C1QG; FLJ27103 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human | |
| 26 kDa | |
| Liquid |
| Immunocytochemistry/Immunofluorescence | |
| 1C5B4 | |
| CoraLite Plus 488 | |
| Mouse | |
| Thiophilic affinity chromatograph | |
| RUO | |
| 714 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction