missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated CD109 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
€ 133.74 - € 361.81
Specifications
| Antigen | CD109 |
|---|---|
| Clone | 2H8G7 |
| Concentration | 1000 μg/mL |
| Applications | Not Available |
| Classification | Monoclonal |
Description
CoraLite™ Plus 488-conjugated CD109 Mouse Monoclonal Antibody, Proteintech shows reactivity in Human samples.Specifications
| CD109 | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 135228 | |
| Peptide | |
| Primary | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 2H8G7 | |
| Not Available | |
| CoraLite Plus 488 | |
| Mouse | |
| Human | |
| CPAMD7; DKFZp762L1111; FLJ38569; FLJ41966; RP11-525G3.1 | |
| IgG1 | |
| Protein G purification | |
| Conjugated Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title