missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated CD40 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66965-100UL
This item is not returnable.
View return policy
Description
The CD40 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD40. This antibody recognizes human antigen. The CD40 antibody has been validated for the following applications: IF-P analysis. This antibody is coralite™ plus 488.Specifications
| CD40 | |
| Monoclonal | |
| 1000 μg/mL | |
| Bp50; CDW40; MGC9013; TNFRSF5; p50 | |
| Recombinant Protein | |
| 100 μL | |
| Primary | |
| Human | |
| Conjugated Antibody | |
| IgG1 |
| Immunofluorescence | |
| 2A8G5 | |
| CoraLite Plus 488 | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 958 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction