missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated Cyclin D1 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-60186-100UL
This item is not returnable.
View return policy
Description
The Cyclin D1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Cyclin D1. This antibody recognizes human, mouse antigen. The Cyclin D1 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.Specifications
| Cyclin D1 | |
| Monoclonal | |
| 1000 μg/mL | |
| BCL1; D11S287E; PRAD1; U21B31 | |
| Recombinant Protein | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| 34 kDa | |
| Liquid |
| Flow Cytometry (Intracellular Staining) | |
| 2G3G5 | |
| CoraLite Plus 488 | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 595 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction