missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated NAPRT1 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66159-20UL
This item is not returnable.
View return policy
Description
The NAPRT1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NAPRT1. This antibody recognizes human, mouse antigen. The NAPRT1 antibody has been validated for the following applications: IF/ICC analysis. This antibody is coralite™ plus 488.Specifications
| NAPRT1 | |
| Monoclonal | |
| 2000 μg/mL | |
| PP3856 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| 51 kDa | |
| Liquid |
| Immunocytochemistry/Immunofluorescence | |
| 5D8H10 | |
| CoraLite Plus 488 | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 93100 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction