missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated EPHA7 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
€ 133.74 - € 361.81
Specifications
| Antigen | EPHA7 |
|---|---|
| Clone | 1A7E5 |
| Concentration | 1000 μg/mL |
| Applications | Not Available |
| Classification | Monoclonal |
Description
CoraLite™ Plus 488-conjugated EPHA7 Mouse Monoclonal Antibody, Proteintech shows reactivity in Human, Mouse, Rat, Pig samples.Specifications
| EPHA7 | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 2045 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 1A7E5 | |
| Not Available | |
| CoraLite Plus 488 | |
| Mouse | |
| Human, Mouse, Rat, Pig | |
| EHK3; HEK11 | |
| IgG1 | |
| Protein G purification | |
| Conjugated Antibody |
Spot an opportunity for improvement?
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.