missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated ERAB Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-60069-100UL
This item is not returnable.
View return policy
Description
CoraLite™ Plus 488-conjugated ERAB Mouse Monoclonal Antibody, Proteintech shows reactivity in Human, Mouse samples.Specifications
| ERAB | |
| Monoclonal | |
| 1000 μg/mL | |
| 17b-HSD10; ABAD; CAMR; DUPXp11.22; ERAB; HADH2; HCD2; MHBD; MRPP2; MRX17; MRX31; MRXS10; SCHAD; SDR5C1 | |
| Recombinant Protein | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| 27 kDa | |
| Liquid | |
| IgG1 |
| Not Available | |
| 1G5E5 | |
| CoraLite Plus 488 | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 3028 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| 25 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction