missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated ESR2 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-60197-20UL
This item is not returnable.
View return policy
Description
The ESR2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ESR2. This antibody recognizes human, mouse antigen. The ESR2 antibody has been validated for the following applications: WB, FC (Intra) analysis. This antibody is coralite™ plus 488.Specifications
| ESR2 | |
| Monoclonal | |
| 1000 μg/mL | |
| ER-BETA; ESR-BETA; ESRB; ESTRB; Erb; NR3A2 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| 50 kDa | |
| Liquid |
| Western Blot, Flow Cytometry (Intracellular Staining) | |
| 5D2C9 | |
| CoraLite Plus 488 | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 2100 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur