missing translation for 'onlineSavingsMsg'
Learn More

CoraLite™ Plus 488-conjugated Glucagon Rabbit Polyclonal Antibody, Proteintech

Product Code. 30236751 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
Unit Size:
20µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30236751 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30236751 Supplier Proteintech Supplier No. CL4881595420UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The Glucagon antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Glucagon. This antibody recognizes human, mouse, rat antigen. The Glucagon antibody has been validated for the following applications: IF-P analysis. This antibody is coralite™ plus 488.

Specifications

Antigen Glucagon
Applications Immunofluorescence
Classification Polyclonal
Concentration 1000 μg/mL
Conjugate CoraLite Plus 488
Gene Symbols GLP1; GLP2; GRPP
Host Species Rabbit
Immunogen Recombinant Protein
Purification Method Antigen affinity purification
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2641
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Product Type Conjugated Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.