missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated GSTO1 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-60150-20UL
This item is not returnable.
View return policy
Description
The GSTO1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GSTO1. This antibody recognizes human, mouse, rat antigen. The GSTO1 antibody has been validated for the following applications: IF/ICC analysis. This antibody is coralite™ plus 488.Specifications
| GSTO1 | |
| Monoclonal | |
| 2000 μg/mL | |
| DKFZp686H13163; GSTTLp28; P28 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 27-30 kDa | |
| Liquid |
| Immunocytochemistry/Immunofluorescence | |
| 5E1E10 | |
| CoraLite Plus 488 | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 9446 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction