missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated IL-1 beta Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
€ 133.74 - € 409.50
Specifications
| Antigen | IL-1 beta |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | Not Available |
| Classification | Polyclonal |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30238022
|
Proteintech
CL488-16806-100UL |
100 μL |
€ 409.50
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30233449
|
Proteintech
CL488-16806-20UL |
20 μL |
€ 133.74
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CoraLite™ Plus 488-conjugated IL-1 beta Rabbit Polyclonal Antibody, Proteintech shows reactivity in Human samples.Specifications
| IL-1 beta | |
| Not Available | |
| CoraLite Plus 488 | |
| Rabbit | |
| Human | |
| IL-1; IL1-BETA; IL1F2 | |
| IgG | |
| Antigen affinity purification | |
| Conjugated Antibody |
| 1000 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 3553 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title