missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated IL-18 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
€ 140.00 - € 450.00
Specifications
| Antigen | IL-18 |
|---|---|
| Clone | 5C6F8 |
| Concentration | 1000 μg/mL |
| Applications | Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30237430
|
Proteintech
CL488-60070-100UL |
100 μL |
€ 450.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30234948
|
Proteintech
CL488-60070-20UL |
20 μL |
€ 140.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The IL-18 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human IL-18. This antibody recognizes human, hamster antigen. The IL-18 antibody has been validated for the following applications: IF/ICC, FC (Intra) analysis. This antibody is coralite™ plus 488.Specifications
| IL-18 | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 3606 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 5C6F8 | |
| Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| CoraLite Plus 488 | |
| Mouse | |
| Human, Hamster | |
| IGIF; IL-18; IL-1g; IL1F4; MGC12320 | |
| IgG2b | |
| Protein A purification | |
| Conjugated Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title