missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated Integrin alpha 6 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
€ 133.74 - € 361.81
Specifications
| Antigen | Integrin alpha 6 |
|---|---|
| Clone | 2B12A1 |
| Concentration | 1000 μg/mL |
| Applications | Not Available |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30234877
|
Proteintech
CL488-66906-100UL |
100 μL |
€ 361.81
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30235776
|
Proteintech
CL488-66906-20UL |
20 μL |
€ 133.74
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CoraLite™ Plus 488-conjugated Integrin alpha 6 Mouse Monoclonal Antibody, Proteintech shows reactivity in Human samples.Specifications
| Integrin alpha 6 | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 3655 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 2B12A1 | |
| Not Available | |
| CoraLite Plus 488 | |
| Mouse | |
| Human | |
| CD49f; DKFZp686J01244; FLJ18737; ITGA6B; VLA-6 | |
| IgG2b | |
| Protein A purification | |
| Conjugated Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title