missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated MAVS; VISA Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-14341-20UL
This item is not returnable.
View return policy
Description
The MAVS; VISA antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MAVS; VISA. This antibody recognizes human antigen. The MAVS; VISA antibody has been validated for the following applications: IF/ICC analysis. This antibody is coralite™ plus 488.Specifications
| MAVS; VISA | |
| Polyclonal | |
| CoraLite Plus 488 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 57506 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgG |
| Immunocytochemistry/Immunofluorescence | |
| 1000 μg/mL | |
| CARDIF; DKFZp547C224; DKFZp666M015; FLJ27482; FLJ35386; FLJ38051; FLJ41962; IPS-1; IPS1; KIAA1271; MGC3260; VISA | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human | |
| 50-55 kDa, 70-75 kDa | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction