missing translation for 'onlineSavingsMsg'
Learn More

CoraLite™ Plus 488-conjugated MEF2C Rabbit Polyclonal Antibody, Proteintech

Product Code. 30234362 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30234362 20 μL 20µL
30237950 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30234362 Supplier Proteintech Supplier No. CL4881005620UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The MEF2C antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MEF2C. This antibody recognizes human, mouse, rat antigen. The MEF2C antibody has been validated for the following applications: IF/ICC analysis. This antibody is coralite™ plus 488.

Specifications

Antigen MEF2C
Applications Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Concentration 1000 μg/mL
Conjugate CoraLite Plus 488
Host Species Rabbit
Immunogen Recombinant Protein
Purification Method Antigen affinity purification
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4208
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Antibody Molecular Weight 52 kDa, 60 kDa
Product Type Conjugated Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.