missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated MEF2C Rabbit Polyclonal Antibody, Proteintech
Shop All Proteintech ProductsDescription
The MEF2C antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MEF2C. This antibody recognizes human, mouse, rat antigen. The MEF2C antibody has been validated for the following applications: IF/ICC analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
| Antigen | MEF2C |
| Applications | Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Concentration | 1000 μg/mL |
| Conjugate | CoraLite Plus 488 |
| Host Species | Rabbit |
| Immunogen | Recombinant Protein |
| Purification Method | Antigen affinity purification |
| Quantity | 20 μL |
| Regulatory Status | RUO |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?