missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated Peripherin Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
€ 140.00 - € 450.00
Specifications
| Antigen | Peripherin |
|---|---|
| Clone | 1H10B3 |
| Concentration | 1000 μg/mL |
| Applications | Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30233769
|
Proteintech
CL488-66317-20UL |
20 μL |
€ 140.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30237620
|
Proteintech
CL488-66317-100UL |
100 μL |
€ 450.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Peripherin antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Peripherin. This antibody recognizes human, rat, pig antigen. The Peripherin antibody has been validated for the following applications: IF/ICC analysis. This antibody is coralite™ plus 488.Specifications
| Peripherin | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 5630 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 1H10B3 | |
| Immunocytochemistry/Immunofluorescence | |
| CoraLite Plus 488 | |
| Mouse | |
| Human, Rat, Pig | |
| NEF4; PRPH1 | |
| IgG2b | |
| Protein A purification | |
| Conjugated Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title