missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 Nucleocapsid Phosphoprotein Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67666-1-IG-20UL
This item is not returnable.
View return policy
Description
The SARS-CoV-2 Nucleocapsid Phosphoprotein antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of virus SARS-CoV-2 Nucleocapsid Phosphoprotein. This antibody recognizes virus antigen. The SARS-CoV-2 Nucleocapsid Phosphoprotein antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| SARS-CoV-2 Nucleocapsid Phosphoprotein | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 43740575 | |
| Store at -20°C. | |
| Liquid |
| Western Blot, ELISA | |
| 1B3C3 | |
| Unconjugated | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Virus | |
| Monoclonal Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction