missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated PTK2B Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-67141-100UL
This item is not returnable.
View return policy
Description
The PTK2B antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PTK2B. This antibody recognizes human, mouse, rat, pig antigen. The PTK2B antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.Specifications
| PTK2B | |
| Monoclonal | |
| 1000 μg/mL | |
| CADTK; CAKB; FADK2; FAK2; FRNK; PKB; PTK; PYK2; RAFTK | |
| Recombinant Protein | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Pig | |
| 112-115 kDa | |
| Liquid |
| Flow Cytometry (Intracellular Staining) | |
| 2A10G5 | |
| CoraLite Plus 488 | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 2185 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction