missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 405 Mouse IgG1 Isotype Control (MOPC-21), Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL405-65124-100UG
This item is not returnable.
View return policy
Description
The IgG1 Isotype Control antibody from Proteintech is a mouse monoclonal antibody generated with n/a of mouse IgG1 Isotype Control. This antibody recognizes antigen. The IgG1 Isotype Control antibody has been validated for the following applications: FC analysis. This antibody is coralite™ plus 405.Specifications
| IgG1 Isotype Control | |
| Monoclonal | |
| 0.2 mg/mL | |
| Mouse | |
| 100 μg | |
| Primary | |
| Not Applicable | |
| Flow Antibody | |
| IgG1 |
| Flow Cytometry | |
| MOPC-21 | |
| CoraLite Plus 405 | |
| Affinity purification | |
| RUO | |
| 16017 | |
| Store at 2-8°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction