missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated Septin 2 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
€ 140.00 - € 450.00
Specifications
| Antigen | Septin 2 |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining) |
| Classification | Polyclonal |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30238136
|
Proteintech
CL488-11397-20UL |
20 μL |
€ 140.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30237531
|
Proteintech
CL488-11397-100UL |
100 μL |
€ 450.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Septin 2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Septin 2. This antibody recognizes human, mouse, rat antigen. The Septin 2 antibody has been validated for the following applications: IF/ICC, FC (Intra) analysis. This antibody is coralite™ plus 488.Specifications
| Septin 2 | |
| Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| CoraLite Plus 488 | |
| Rabbit | |
| Human, Mouse, Rat | |
| DIFF6; KIAA0158; NEDD5; Pnutl3; hNedd5 | |
| IgG | |
| Antigen affinity purification | |
| Conjugated Antibody |
| 1000 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 4735 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title