missing translation for 'onlineSavingsMsg'
Learn More

CoraLite™ Plus 488-conjugated TPT1 Mouse Monoclonal Antibody, Proteintech

Product Code. 30235799 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30235799 100 μL 100µL
30237467 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30235799 Supplier Proteintech Supplier No. CL48866713100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The TPT1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TPT1. This antibody recognizes human, mouse, rat antigen. The TPT1 antibody has been validated for the following applications: IF/ICC analysis. This antibody is coralite™ plus 488.

Specifications

Antigen TPT1
Applications Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone 1G6A10
Concentration 1000 μg/mL
Conjugate CoraLite Plus 488
Gene Symbols FLJ27337; HRF; TCTP; p02
Host Species Mouse
Immunogen Recombinant Protein
Purification Method Protein G purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7178
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Antibody Molecular Weight 22 kDa
Product Type Conjugated Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.