missing translation for 'onlineSavingsMsg'
Learn More

CoraLite™ Plus 488-conjugated Zinc Alpha 2 Glycoprotein Mouse Monoclonal Antibody, Proteintech

Mouse Monoclonal Antibody

€ 140.00 - € 450.00

Specifications

Antigen Zinc Alpha 2 Glycoprotein
Clone 4B8A4
Concentration 1000 μg/mL
Applications Immunocytochemistry/Immunofluorescence
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30234494
missing translation for 'viewDocuments'
Proteintech
CL488-66178-100UL
100 μL
€ 450.00
100µL
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
30237334
missing translation for 'viewDocuments'
Proteintech
CL488-66178-20UL
20 μL
€ 140.00
20µL
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Zinc Alpha 2 Glycoprotein antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Zinc Alpha 2 Glycoprotein. This antibody recognizes human antigen. The Zinc Alpha 2 Glycoprotein antibody has been validated for the following applications: IF/ICC analysis. This antibody is coralite™ plus 488.
Specifications

Specifications

Zinc Alpha 2 Glycoprotein
1000 μg/mL
Monoclonal
Liquid
RUO
563
Recombinant Protein
Primary
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
4B8A4
Immunocytochemistry/Immunofluorescence
CoraLite Plus 488
Mouse
Human
ZA2G; ZAG
IgG1
Protein G purification
Conjugated Antibody
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.