missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CORIN Polyclonal Antibody

Product Code. 16566241
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16566241 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16566241 Supplier Invitrogen™ Supplier No. PA587872

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MKQSPALAPE ERCRRAGSPK PVLRADDNNM GNGCSQKLAT ANLLRFLLLV LIPCICALVL LLVILLSYVG TLQKVYFKSN GSEPLVTDGE IQGSDVILTN TIYNQSTVVS; Positive Samples: Hela, HT-1080, Mouse heart, Mouse brain, Rat heart, Rat brain; Cellular Location: Cell membrane, Single-pass type II membrane protein, Single-pass type II membrane protein.

CORIN is a member of the type II transmembrane serine protease class of the trypsin superfamily. Members of this family are composed of multiple structurally distinct domains. CORIN converts pro-atrial natriuretic peptide to biologically active atrial natriuretic peptide, a cardiac hormone that regulates blood volume and pressure. This protein may also function as a pro-brain-type natriuretic peptide convertase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CORIN
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.99 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CORIN
Gene Accession No. Q80YN4, Q9Y5Q5, Q9Z319
Gene Alias ATC2; Atrial natriuretic peptide-converting enzyme; Atrial natriuretic peptide-converting enzyme, 100 kDa soluble fragment; Atrial natriuretic peptide-converting enzyme, 160 kDa soluble fragment; Atrial natriuretic peptide-converting enzyme, 180 kDa soluble fragment; Atrial natriuretic peptide-converting enzyme, activated protease fragment; Atrial natriuretic peptide-converting enzyme, N-terminal propeptide; AV273130; Corin; corin, serine peptidase; CRN; EC 3.4.21; Heart-specific serine proteinase ATC2; low density lipoprotein receptor-related protein 4; low density lipoprotein-related protein 4; Lrp4; MGC119742; PEE5; pro-ANP-convertase; pro-ANP-converting enzyme; PRSC; serine peptidase; serine protease; TMPRSS10; Transmembrane protease serine 10
Gene Symbols CORIN
Host Species Rabbit
Immunogen Recombinant protein (or fragments).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10699, 289596, 53419
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.