Learn More
JULABO™ CORIO™ CP-601F Refrigerated/Heating Circulator
Product Code. 16499339
Change view
Click to view available options
Capacity (Metric):
8 to 10 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
16499339 8 to 10 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16499339 Supplier JULABO™ Supplier No. 9013705
Refrigerated Circulators feature enhanced pump performance for easy temperature control tasks up to 200°C.
  • Models for internal and external applications
  • Bright, white, easy to read display
  • Very quiet
  • Easy switching between internal and external circulation
  • External pump connections
  • Powerful and infinitely adjustable pressure pump
  • USB connection
  • RS232 interface for online communication
  • Space-saving cooling coil design yields more usable space in the bath tank
  • Bath lid and drain tap included
  • Removable ventilation grid
  • Refrigeration unit without side vents
  • Class III (FL) according to DIN 12876-1
Specifications
Ambient Temperature 5 to 40°C
Controller Type PID1
Capacity (Metric) 8 to 10 L
Cooling Capacity 0.6 kW at 20°C
Flow Rate 8 to 27 L/min.
Temperature Range -35°C to +200°C
Display LED
External Circulation Yes
Heating Capacity 2 kW
Includes 2 Barbed fittings for tubing 8 and 12 mm ID. (Pump connections M16x1 male)
Pump Capacity 0.1 to 0.7 bar
Refrigerant R449A
Temperature Stability ±0.03°C
Timer Yes
USB Connections Yes
Viscosity Max. 50 cST
Dimensions (D x W x H) Interior 22 x 15 x 20 cm
Dimensions (L x W x H) 46 x 36 x 74 cm
Weight (Metric) 38.5 kg