Learn More
JULABO™ CORIO™ CP-BC26 Heating Circulator
Product Code. 16459339
Change view
Click to view available options
Capacity (Metric):
19 to 26 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
16459339 19 to 26 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16459339 Supplier JULABO™ Supplier No. 9013526
CORIO™ Circulator can be equipped with an optional pump set for temperature control of external applications.
  • Models for internal and external applications
  • Bright, white, easy to read display
  • Very quiet
  • Easy switching between internal and external circulation
  • Powerful and infinitely adjustable pressure pump
  • USB connection
  • RS232 interface for online communication
  • High-quality bath tanks made of stainless steel with bath lid and drain tap
  • Integrated pump connection M16 x 1
  • Integrated cooling coil for counter-cooling
  • Precise temperature control
Specifications
Ambient Temperature 5 to 40°C
Controller Type PID1
Capacity (Metric) 19 to 26 L
Flow Rate 8 to 27 L/min.
Temperature Range 20°C to 200°C
Display LED
External Circulation Yes
Heating Capacity 2 kW
Includes 2 Barbed fittings for tubing 8 and 12 mm ID. (Pump connections M16x1 male)
Pump Capacity 0.1 to 0.7 bar
Temperature Stability ±0.02°C
Timer Yes
USB Connections Yes
Viscosity Max. 50 cST
Dimensions (D x W x H) Interior 26 x 35 x 20 cm
Dimensions (L x W x H) 62 x 39 x 48 cm
Weight (Metric) 19 kg