Learn More
Corn Oil, TRC
Click to view available options
Quantity:
100 mg
Conditionnement:
100mg
Spécification
Spécification
| Chemical Name or Material | Corn Oil |
| CAS | 8001-30-7 |
| Quantity | 100 mg |
| Synonym | Bunge, Corn Germ Oil, Germ Oil of Semen Maydis, Gold-N-Sweet Pan Spray, Lipomul, Maydol, Maise Oil, Mazola Oil, Semen Maydis Oil, Semen Maydis Salad Oil, Uni Ace CS |