missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 1536-Well Black Clear-Bottom Polystyrene Plates

Product Code. 10172912 Shop All Corning Products
Click to view available options
Color:
Black, Clear Bottom
Unit Size:
Pack of 50
This item is not returnable. View return policy

Product Code. 10172912

Brand: Corning™ 3891

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Highest density microplates available for high throughput screening

Corning 1536-well microplates are our highest density microplates available for high throughput screening. The microplates conform to standard microplate footprint and dimensions. These microplates are offered in solid black and white polystyrene, with round or flat bottoms, and in black-clear bottom formats.

  • Total well volume of 10 μL for round well microplates and 12.8 μL for flat bottom microplates
  • Recommended working volume up to 8 μL
  • Round well bottom for reduced air entrapment and improved CV values and Z factor
  • Raised well bottom for higher sensitivity
  • Flood reservoir on four sides to reduce instrument contamination
  • Lids are available separately. Corning lid (Cat. No. 3098) is compatible with these microplates.

Order Info

Packaged in bags of 10

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Lid Without Lid
Array 32 x 48
Packaging 50/Pk.
Color Black, Clear Bottom
Product Type 1536-Well Assay Microplate
For Use With (Equipment) Automated dispensers
Material Polystyrene
Volume (Metric) Well 12.8 μL
No. of Wells 1536
Volume (Metric) Working 8 μL
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Flat Bottom (F-well)
For Use With (Application) For fluorescent homogeneous assays, high throughput screening
No. per Case 50/Cs.
No. per Pack 10/Pk.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.