Learn More
Corning™ BioCoat™ 384-Well, Poly-D Lysine-Treated, Flat-Bottom Microplate
Color:Black, Clear Bottom; Sterility:Non-sterile; Lid:With Lid; Type: Optical Glass Base
Brand: Corning™ 4587
Additional Details : Weight : 3.28000kg
Description
High optical quality, glass bottom black microplates are ideal for performing high-content cell-based assays using imaging systems. The glass bottom provides a flat and optically clear surface that reduces autofocus time, increases throughput, and is ideal for cell growth.
- High optical quality and scratch resistant glass
- Glass bottom thickness of 200 μm is well suited for imaging microscopy.
- Bottom flatness <50 μm to ensure planarity for imaging devices
- Low background fluorescence and minimal cross-talk provide the highest possible optical quality for cell-based assays.
Specifications
Half-Area | |
16 x 24 | |
Black, Clear Bottom | |
384 | |
Poly-D-Lysine | |
10/Cs. |
With Lid | |
1/Pk., 10/Cs. | |
Optical Glass Base | |
Non-sterile | |
Flat Bottom (F-well) | |
1/Pk. |