Learn More
Corning™ BioCoat™ TC-treated Cultureslide Variety Pack
Click to view available options
No. of Wells:
8
Unit Size:
Pack of 12
Description
- Selections include different combinations of extracellular matrix proteins and attachment factors
Cultureslide Variety Packs
- 8-well
- Pack contains twelve slides coated with Collagen I, Fibronectic and Poly-D-Lysine
Specifications
Specifications
| Type | Culture Slide |
| Storage Requirements | 2° to 8°C |
| No. of Wells | 8 |